Learn More
Thermo Scientific™ Dionex™ IonPac™ NS1 Analytical & Guard Columns
Achieve high-efficiency separations of hydrophobic, ionizable compounds with these polymeric-based, reversed-phase columns.
Brand: Thermo Scientific™ 035321
740.62 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
Achieve high-efficiency separations of hydrophobic, ionizable compounds with Thermo Scientific™ Dionex™ IonPac™ NS1 10μm Analytical & Guard Columns. The columns of choice for ion-pair or ion-suppression chromatography, these polymer-based, reversed-phase columns provide excellent resolution and peak shape. Packing is a neutral, macroporous, high surface area, ethylvinylbenzene polymer cross linked with 55% divinylbenzene, making IonPac NS1 columns resistant to solvents, acids and bases while permitting the use of eluent from pH 0 to 14.
Polymer-Based Columns for Efficient Reversed-Phase Separations- Ideal for separation of large molecules that carry localized charges, such as surfactants
- Can be used for traditional polymeric reversed-phase applications
- Use with ion-pair chromatography for difficult separations
- Available in 5 and 10μm particle sizes
Use IonPac NS1 columns for reliable separations of various compounds: High molecular weight aliphatic, carboxylic acids; Alkyl and aromatic sulfates and sulfonates; Quaternary ammonium ions; Water-soluble vitamins; Sulfur oxides; Metal cyanide complexes; Phenols
Specifications
Mixed Mode | |
1 mL/min | |
High | |
10 μm | |
Spherical, Crosslinked, Macroporous Polymer | |
Each | |
250 mm |
55%DVB | |
Reversed-Phase or Ion-Pair Chromatography | |
4000 psi (275 bar) | |
Ethylvinylbenzene/Divinylbenzene | |
0 to 14 | |
0 to 100% of HPLC solvents | |
4 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.