missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Dionex™ IonPac™ NS1 and NG1 IC Columns

Product Code. p-4524123 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
35 mm
50 mm
150 mm
250 mm
Diameter (Metric):
2 mm
4 mm
Particle Size:
5 μm
10 μm
Column Type:
Analytical
Guard
Mixed Mode
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11391693

Brand: Thermo Scientific™ 035321

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Achieve high-efficiency separations of hydrophobic, ionizable compounds with these polymeric-based, reversed-phase columns.

Achieve high-efficiency separations of hydrophobic, ionizable compounds with Thermo Scientific™ Dionex™ IonPac™ NS1 10μm Analytical & Guard Columns. The columns of choice for ion-pair or ion-suppression chromatography, these polymer-based, reversed-phase columns provide excellent resolution and peak shape. Packing is a neutral, macroporous, high surface area, ethylvinylbenzene polymer cross linked with 55% divinylbenzene, making IonPac NS1 columns resistant to solvents, acids and bases while permitting the use of eluent from pH 0 to 14.

Polymer-Based Columns for Efficient Reversed-Phase Separations
  • Ideal for separation of large molecules that carry localized charges, such as surfactants
  • Can be used for traditional polymeric reversed-phase applications
  • Use with ion-pair chromatography for difficult separations
  • Available in 5 and 10μm particle sizes
Key Applications
Use IonPac NS1 columns for reliable separations of various compounds: High molecular weight aliphatic, carboxylic acids; Alkyl and aromatic sulfates and sulfonates; Quaternary ammonium ions; Water-soluble vitamins; Sulfur oxides; Metal cyanide complexes; Phenols
TRUSTED_SUSTAINABILITY

Specifications

Column Type Analytical
Crosslinking 55%DVB
Flow Rate 1 mL/min
For Use With (Application) Reversed-Phase or Ion-Pair Chromatography
Hydrophobicity High
Max. Pressure 4000 psi (275 bar)
Particle Size 10 μm
Stationary Phase Ethylvinylbenzene/Divinylbenzene
Type Mixed Mode
Format Standard Bore
Packing Material Spherical, Crosslinked, Macroporous Polymer
pH 0 to 14
Solvents 0 to 100% of HPLC solvents
Length (Metric) 250 mm
Diameter (Metric) 4 mm
Green Features Responsibly packaged
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.