Learn More
Thermo Scientific™ Dionex™ MAbPac™SCX-10 Ion Exchange Columns
For the high-resolution separation and analysis of monoclonal antibody variants
Brand:  Thermo Scientific™ 088783
 View more versions of this product
Product Code. 15530244
  • £3381.83 / Each
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Unequalled high-resolution separations
  • Exceptionally high efficiency
  • Highest resolution of monoclonal antibody variants
  • Unmatched column-to-column and lot-to-lot reproducibility
  • Hydrophobic interactions eliminated
  • Excellent for stability testing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bio-LC
3 Lots of Resin