Learn More
Thermo Scientific™ OmniPac™ PCX-500 HPLC Columns
Separate cationic and neutral species in a single run using this solvent-compatible mixed-mode column.
£501.50 - £1331.63
Specifications
Column Type | Mixed Mode |
---|---|
For Use With (Application) | Reversed Phase and Ion Exchange |
Max. Pressure | 4000 psi (275 bar) |
Particle Size | 8.5 μm |
Stationary Phase | Sulfonic Acid |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
11351723
|
Thermo Scientific™
042195 |
50 mm |
£501.50
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
11331723
|
Thermo Scientific™
042191 |
250 mm |
£1331.63
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
Description
Separate cationic and neutral analytes in a single run by using Thermo Scientific™ OmniPac™ PCX-500 Columns. With cation-exchange and reversed-phase properties, this polymer-based mixed-mode column is solvent compatible and stable over the entire pH range of 1-14. Solvents may be used to affect selectivity as well as to remove contaminants to extend column lifetime.
This column is ideal for analyzing a wide range of compounds including adrenergics, alcohols, anilines, antidepressants, anti-inflammatory agents, plating bath brighteners, cephalosporins, stains and dyes, herbicides, nucleotides and nucleosides, peptides, sulfonamides, and water-soluble vitamins.- Macroporous hydrophobic core provides reversed-phase retention and is coated with an cation-exchange latex bead layer for cation-exchange retention
- Ion exchange capacity of approximately 120 μeq/column
- 100% solvent-compatible and should always be run with at least 1% organic solvent in the mobile phase
Specifications
Mixed Mode | |
4000 psi (275 bar) | |
Sulfonic Acid | |
0 to 14 | |
Each | |
4 mm |
Reversed Phase and Ion Exchange | |
8.5 μm | |
Spherical, Microporous Polymer | |
1200 Å | |
L50 | |
Responsibly packaged |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.