Learn More
Thermo Scientific™ Dionex™ Specially Formulated PTFE Tubing
Product Code. p-6082207 Shop All Thermo Scientific Products
Click to view available options
Description:
250 and 350 um ID
250 um ID; Low pressure
350 um ID; Low pressure
For Use With (Equipment):
Dionex UltiMate 3000 RSLC Nano System
Fused Silica Capillary Tubing
Unit Size:
Pack of 2
Pack of 5
Product Code. 11322133
Brand: Thermo Scientific™ 160486
Only null left
Add to basket
Add to basket
Thermo Scientific™ Dionex Specially Formulated PTFE Tubing is used as connector for fused silica tubing (nano and cap LC). Choose fitting dimensions for OD of fused silica tubing used.
Thermo Scientific™ Dionex Specially Formulated PTFE Tubing is used as connector for fused silica tubing (nano and cap LC).
TRUSTED_SUSTAINABILITY
Specifications
Description 250 um ID; Low pressure
For Use With (Equipment) Dionex UltiMate 3000 RSLC Nano System
Type Capillary Tubing
Product Line nanoLC