Learn More
Thermo Scientific™ Dionex™ Ultra-Low Dead Volume Union
Product Code. 11392133 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Nano ES
missing translation for 'unitSize'
Each
Product Code. 11392133
missing translation for 'mfr': Thermo Scientific™ 160494
Only null left
Add to basket
Add to basket
Thermo Scientific™ Ultra-Low Dead Volume Union is for nano electrospray ionization.
Thermo Scientific™ Ultra-Low Dead Volume Union is for nano electrospray ionization.
  • Dead volume of <7nL
  • With two nuts and two ferrules
  • I.D.: 1/16in.
TRUSTED_SUSTAINABILITY
Specifications
Description Union
For Use With (Equipment) Nano ES
Type Union
Diameter (English) Outer 1/16 in.
Product Line nanoLC