missing translation for 'onlineSavingsMsg'
Learn More

Dipentene, pure, fraction of terpene hydrocarbons, Thermo Scientific Chemicals

Product Code. 10398403
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68956-56-9
Molecular Formula C10H16
Molecular Weight (g/mol) 136.23
MDL Number MFCD00062992
Synonym spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl

Specifications

Density 0.8680g/mL
Boiling Point 174.0°C
Flash Point 42°C
Packaging Glass bottle
Refractive Index 1.4730 to 1.4780 (20°C, 589nm)
Quantity 1 L
Specific Gravity 0.868
Formula Weight 136.23
Grade Pure
Physical Form Liquid
Chemical Name or Material Dipentene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dipentene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'