missing translation for 'onlineSavingsMsg'
Learn More

Dipentyl phthalate, 97%, Thermo Scientific Chemicals

Product Code. 10276051
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-18-0
Molecular Formula C18H26O4
Molecular Weight (g/mol) 306.4
InChI Key IPKKHRVROFYTEK-UHFFFAOYSA-N
Synonym dipentyl phthalate, di-n-pentyl phthalate, diamyl phthalate, amyl phthalate, amoil, di-n-amyl phthalate, di-n-pentylphthalate, phthalic acid, dipentyl ester, phthalic acid diamyl ester, 1,2-benzenedicarboxylic acid, dipentyl ester
PubChem CID 8561
ChEBI CHEBI:34680
IUPAC Name dipentyl benzene-1,2-dicarboxylate
SMILES CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC

Specifications

Melting Point -55.0°C
Color Colorless to Yellow
Density 1.0230g/mL
Boiling Point 342.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C(O)OC5H11)2C6H4
Refractive Index 1.4885 to 1.4905
Quantity 100 g
Specific Gravity 1.023
Formula Weight 306.4
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dipentyl phthalate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dipentyl phthalate
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1B
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.