missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl phosphate, 97%

Product Code. p-7043693
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11361517

Brand: Thermo Scientific Alfa Aesar L02121.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 838-85-7 | C12H11O4P | 250.19 g/mol

Diphenyl phosphate is used in the manufacture of phosphoric acid diesters such as triphenyl phosphate, trixylenyl phosphate, isodecyl diphenyl phosphate, cresyl diphenyl phosphate and isopropylphenyl diphenyl phosphate. It is widely used as a protective agent for hydroxyl group in organic synthesis. It finds application as an additive for paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl phosphate is used in the manufacture of phosphoric acid diesters such as triphenyl phosphate, trixylenyl phosphate, isodecyl diphenyl phosphate, cresyl diphenyl phosphate and isopropylphenyl diphenyl phosphate. It is widely used as a protective agent for hydroxyl group in organic synthesis. It finds application as an additive for paints and coatings.

Solubility
Soluble in benzene. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 838-85-7
Molecular Formula C12H11O4P
Molecular Weight (g/mol) 250.19
MDL Number MFCD00003033
InChI Key ASMQGLCHMVWBQR-UHFFFAOYSA-N
Synonym diphenyl phosphate, phosphoric acid, diphenyl ester, phenyl hydrogen phosphate, phenyl phosphate pho 2 ho po, diphenoxyphosphinic acid, phosphoric acid diphenyl ester, diphenylphosphoric acid, phosphoric acid diphenyl, dsstox_cid_28182
PubChem CID 13282
IUPAC Name diphenyl hydrogen phosphate
SMILES OP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Melting Point 62°C to 70°C
Quantity 5 g
Beilstein 1379164
Solubility Information Soluble in benzene. Insoluble in water.
Formula Weight 250.19
Percent Purity 97%
Chemical Name or Material Diphenyl phosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.