missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl phosphoramidate, 97%

Product Code. 11347156
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11347156

Brand: Thermo Scientific Alfa Aesar L07155.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2015-56-7 | C12H12NO3P | 249.206 g/mol

Diphenyl phosphoramidate is used as primary and secondary intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl phosphoramidate is used as primary and secondary intermediates.

Solubility
Insoluble in water.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2015-56-7
Molecular Formula C12H12NO3P
Molecular Weight (g/mol) 249.206
MDL Number MFCD00014078
InChI Key QWMUDOFWQWBHFI-UHFFFAOYSA-N
Synonym diphenyl phosphoramidate, diphenyl amidophosphate, phosphoramidic acid, diphenyl ester, diphenylphosphoramidate, amino phenoxy phosphoryl oxy benzene, diphenyl phosphoroamidate, aminodiphenoxyphosphino-1-one, amidophosphoric acid, diphenyl ester, acmc-1cki3
PubChem CID 74836
IUPAC Name [amino(phenoxy)phosphoryl]oxybenzene
SMILES C1=CC=C(C=C1)OP(=O)(N)OC2=CC=CC=C2

Specifications

Melting Point 147°C to 151°C
Quantity 5 g
Beilstein 2054166
Solubility Information Insoluble in water.
Formula Weight 249.21
Percent Purity 97%
Chemical Name or Material Diphenyl phosphoramidate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.