missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl phthalate, 98%, Thermo Scientific Chemicals

Product Code. 11467107
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-62-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00003038
InChI Key DWNAQMUDCDVSLT-UHFFFAOYSA-N
Synonym diphenyl phthalate, phenyl phthalate, phthalic acid, diphenyl ester, diphenylphthalate, 1,2-benzenedicarboxylic acid, diphenyl ester, phthalic acid diphenyl ester, caswell no. 399b, unii-bu20109xhv, 1,2-benzenedicarboxylic acid, 1,2-diphenyl ester, epa pesticide chemical code 017001
PubChem CID 6778
ChEBI CHEBI:60819
IUPAC Name diphenyl benzene-1,2-dicarboxylate
SMILES O=C(OC1=CC=CC=C1)C1=CC=CC=C1C(=O)OC1=CC=CC=C1

Specifications

Melting Point 74°C to 76°C
Density 1.28
Boiling Point 400°C to 405°C
Flash Point 224°C (435°F)
Odor Odorless
Quantity 100 g
Beilstein 2473390
Merck Index 14,7306
Formula Weight 318.33
Percent Purity 98%
Chemical Name or Material Diphenyl phthalate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'