Learn More
Chemical Identifiers
CAS | 127-63-9 |
---|---|
Molecular Formula | C12H10O2S |
Molecular Weight (g/mol) | 218.27 |
MDL Number | MFCD00007548 |
InChI Key | KZTYYGOKRVBIMI-UHFFFAOYSA-N |
Synonym | diphenyl sulfone, phenyl sulfone, sulfonyldibenzene, diphenylsulfone, sulfobenzide, diphenyl sulphone, benzene, 1,1'-sulfonylbis, sulfone, diphenyl, 1,1'-sulfonyldibenzene, phenyl sulphone |
PubChem CID | 31386 |
ChEBI | CHEBI:78360 |
IUPAC Name | benzenesulfonylbenzene |
SMILES | O=S(=O)(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11338055
|
Thermo Scientific Alfa Aesar
L08232.30 |
250 g |
£29.55
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11348055
|
Thermo Scientific Alfa Aesar
|
1000 g |
£85.63
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a high temperature solvent. Such high temperature solvents are useful for processing highly rigid polymers, e.g., PEEK, which only dissolve in very hot solvents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a high temperature solvent. Such high temperature solvents are useful for processing highly rigid polymers, e.g., PEEK, which only dissolve in very hot solvents.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
127-63-9 | |
218.27 | |
KZTYYGOKRVBIMI-UHFFFAOYSA-N | |
31386 | |
benzenesulfonylbenzene |
C12H10O2S | |
MFCD00007548 | |
diphenyl sulfone, phenyl sulfone, sulfonyldibenzene, diphenylsulfone, sulfobenzide, diphenyl sulphone, benzene, 1,1'-sulfonylbis, sulfone, diphenyl, 1,1'-sulfonyldibenzene, phenyl sulphone | |
CHEBI:78360 | |
O=S(=O)(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
127-63-9 | |
1.36 | |
184°C (363°F) | |
C12H10O2S | |
1910573 | |
diphenyl sulfone, phenyl sulfone, sulfonyldibenzene, diphenylsulfone, sulfobenzide, diphenyl sulphone, benzene, 1,1'-sulfonylbis, sulfone, diphenyl, 1,1'-sulfonyldibenzene, phenyl sulphone | |
KZTYYGOKRVBIMI-UHFFFAOYSA-N | |
benzenesulfonylbenzene | |
31386 | |
218.27 | |
Diphenyl sulfone |
123°C to 130°C | |
377°C to 379°C | |
Odorless | |
MFCD00007548 | |
14,3238 | |
Insoluble in water. | |
O=S(=O)(C1=CC=CC=C1)C1=CC=CC=C1 | |
218.27 | |
CHEBI:78360 | |
≥99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 204-853-1
missing translation for 'rtecsNumber' : SX2400000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Diphenyl sulfone
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Diphenylsulfone
RUO – Research Use Only