missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylamine-4-sulfonic acid, sodium salt, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
Color | Beige to White or Gray |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Linear Formula | C6H5NHC6H4SO3Na |
Quantity | 1 g |
Beilstein | 14,7 |
Sensitivity | (as indicator) Passes Test |
Merck Index | 15,7423 |
Solubility Information | Solubility in water: soluble. |
Formula Weight | 271.26 |
Show More |
Safety and Handling
Product Identifier
- Diphenylamine-4-sulfonic acid, sodium salt
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction