missing translation for 'onlineSavingsMsg'
Learn More

Diphenylamine R1 solution EU Pharmacopoeia, Fisher Chemical™

Product Code. 12980404
Click to view available options
Quantity:
1000 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

CAS 6988-21-2
Packaging Glass bottle
Quantity 1000 mL
Molecular Formula C11H13NO4
InChI Key SDKQRNRRDYRQKY-UHFFFAOYSA-N
SMILES CNC(=O)OC1=CC=CC=C1C2OCCO2
IUPAC Name [2-(1,3-dioxolan-2-yl)phenyl] N-methylcarbamate
Molecular Weight (g/mol) 223.228
PubChem CID 23421
ChEBI CHEBI:82093
Grade EU Ph Ref: 1032102
Analyte or Component Names Diphenylamine solution R1.
Solvent or Matrix 6988-21-2
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • diphenyl amine solution R1
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands, face and anyexposed skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : diphenyl amine, sulphuric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'