missing translation for 'onlineSavingsMsg'
Learn More

Diphenylamine Solution, Reagecon™
BIOPHARMA

Product Code. 12683623
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Manufactured and tested in compliance with the Chapter 4.1.1 of the European Pharmacopoeia.

Supplied with a Certificates of Analysis. Lot number and expiry date are stated on all product labels.

  • Synonyms: DPA
  • Molecular Formula: (C6H5)2NH
  • Molar Mass: 169.23g/mol
  • CAS: 122-39-4

Specifications

Color Colorless
Density 1.83921g/cm3
Boiling Point 295°C
Molecular Formula (C6H5)2NH
Quantity 1 L
Solubility Information Fully miscible
Formula Weight 169.23g/mol
Concentration or Composition (by Analyte or Components) ≤2.5% DPA in Sulphuric Acid
Physical Form Liquid
compliance-icons
Product Identifier
  • DIPHENYLAMINE SOLUTION
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, diphenylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'