missing translation for 'onlineSavingsMsg'
Learn More

Diphenylborinic acid 2-aminoethyl ester, 98%

Product Code. 10552171 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
5g
This item is not returnable. View return policy

Product Code. 10552171

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 524-95-8 | C14H16BNO | 225.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 524-95-8
Molecular Formula C14H16BNO
Molecular Weight (g/mol) 225.10
MDL Number MFCD00014823
InChI Key BLZVCIGGICSWIG-UHFFFAOYSA-N
Synonym 2-aminoethyl diphenylborinate, 2-apb, 2-aminoethoxy diphenylborane, 2-aminoethoxydiphenylborane, 2-aminoethoxydiphenylborate, 2-aminoethoxydiphenyl borate, o-2-aminoethyl diphenylborinic acid, b-2-aminoethoxy diphenylborane, 2-diphenylboryl oxy ethanamine, borinic acid, o-2-aminoethyl diphenyl
PubChem CID 1598
SMILES NCCOB(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 190.0°C to 194.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C6H5)2BOCH2CH2NH2
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,methanol
IUPAC Name 2-diphenylboranyloxyethanamine
Formula Weight 225.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diphenylborinic acid 2-aminoethyl ester, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Diphenylborinic acid 2-aminoethyl ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.