missing translation for 'onlineSavingsMsg'
Learn More
Diphenylcyclopropenone, 98%, Thermo Scientific Chemicals Product Code.: 10317740

Diphenylcyclopropenone, 98%, Thermo Scientific Chemicals

Product Code. 10317740
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10317740

Brand: Thermo Scientific Acros 175820050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886-38-4
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.24
MDL Number MFCD00001311
InChI Key HCIBTBXNLVOFER-UHFFFAOYSA-N
Synonym diphenylcyclopropenone, diphencyprone, 2,3-diphenylcyclopropenone, 2,3-diphenylcycloprop-2-enone, cyclopropenone, diphenyl, dpcp, 2-cyclopropen-1-one, 2,3-diphenyl, 1,2-diphenylcyclopropen-3-one, cyclopropenone, 2,3-diphenyl, unii-i7g14nw5ec
PubChem CID 65057
ChEBI CHEBI:53074
IUPAC Name 2,3-diphenylcycloprop-2-en-1-one
SMILES O=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 117.0°C to 121.0°C
Color Beige to Orange-Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 07,IV,1736
Merck Index 15,3338
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol, ethanol, acetone, chloroform,, dichloromethane, cyclohexane, benzene, hexane,, petroleum ether
Formula Weight 206.24
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Diphenylcyclopropenone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diphenylcyclopropenone, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.