missing translation for 'onlineSavingsMsg'
Learn More

Diphenyldichlorosilane, 97%, Thermo Scientific Chemicals

£31.36 - £105.53

Chemical Identifiers

CAS 80-10-4
Molecular Formula C12H10Cl2Si
Molecular Weight (g/mol) 253.20
MDL Number MFCD00000489
InChI Key OSXYHAQZDCICNX-UHFFFAOYSA-N
Synonym diphenyldichlorosilane, silane, dichlorodiphenyl, dichloro diphenyl silane, diphenylsilicon dichloride, diphenylsilyl dichloride, dichlor-difenylsilan, diphenyl dichlorosilane, dichlor-difenylsilan czech, hsdb 316, benzene, 1,1'-dichlorosilylene bis
PubChem CID 6627
IUPAC Name dichloro(diphenyl)silane
SMILES Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11419932
View Documents
Thermo Scientific Alfa Aesar
A12051.22
100 g
£31.36
100g
Please to purchase this item. Need a web account? Register with us today!
11429932
View Documents
Thermo Scientific Alfa Aesar
A12051.36
500 g
£105.53
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuffs.

Solubility
Decomposes in water.

Notes
Moisture Sensitive. Store away from water/moisture, oxidizing agents and bases. Store under inert gas.
Specifications

Chemical Identifiers

80-10-4
253.20
OSXYHAQZDCICNX-UHFFFAOYSA-N
6627
Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1
C12H10Cl2Si
MFCD00000489
diphenyldichlorosilane, silane, dichlorodiphenyl, dichloro diphenyl silane, diphenylsilicon dichloride, diphenylsilyl dichloride, dichlor-difenylsilan, diphenyl dichlorosilane, dichlor-difenylsilan czech, hsdb 316, benzene, 1,1'-dichlorosilylene bis
dichloro(diphenyl)silane

Specifications

303°C to 304°C
80-10-4
MFCD00000489
609882
diphenyldichlorosilane, silane, dichlorodiphenyl, dichloro diphenyl silane, diphenylsilicon dichloride, diphenylsilyl dichloride, dichlor-difenylsilan, diphenyl dichlorosilane, dichlor-difenylsilan czech, hsdb 316, benzene, 1,1'-dichlorosilylene bis
OSXYHAQZDCICNX-UHFFFAOYSA-N
dichloro(diphenyl)silane
6627
97%
1.579
1.208
-22°C
C12H10Cl2Si
UN1769
157°C (314°F)
Decomposes in water.
Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1
253.20
253.2
Pungent
Moisture sensitive
Diphenyldichlorosilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H310-H314-H318-H302
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H310-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DIPHENYLDICHLOROSILANE

missing translation for 'einecsNumber' : 201-251-0

missing translation for 'rtecsNumber' : VV3190000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Product Identifier
  • Diphenyldichlorosilane

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Diphenyldichlorosilane

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only