missing translation for 'onlineSavingsMsg'
Learn More
Diphenyleneiodonium chloride, Tocris Bioscience™ Product Code.: 18014162

Diphenyleneiodonium chloride, Tocris Bioscience™

Product Code. 18014162
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18014162

Brand: Tocris Bioscience 0504/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GPR3 agonist; also inhibits NOS and NADPH oxidases

GPR3 agonist (EC50 = 1 μM); activates adenylate cyclase through GPR3 but not GPR6 or GPR12. Also induces Ca2+ mobilization and β-arrestin receptor internalization. Binds strongly to flavoproteins; inhibits several enzymes, including NO synthase, NADPH oxidases and NADPH cytochrome P450 oxidoreductase. Also inhibits platelet aggregation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4673-26-1
Molecular Formula C12H8ClI
Molecular Weight (g/mol) 314.55
InChI Key FCFZKAVCDNTYID-UHFFFAOYSA-M
Synonym diphenyleneiodonium chloride, dibenziodolium chloride, unii-7m9d81yz2n, dpi, 8? 3-iodatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2,4,6,9,11-hexaen-8-ylium chloride, dibenziodolium, chloride, d0n0gj, c12h8i.cl, dibenzo b,d iodolium chloride, 2,2'-biphenylyleneiodonium chloride
PubChem CID 2733504
ChEBI CHEBI:77967
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3[I+]2.[Cl-]

Specifications

Quantity 10 mg
Formula Weight 314.55
Percent Purity >99%
Chemical Name or Material Diphenyleneiodonium chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diphenyleneiodonium chloride, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.