missing translation for 'onlineSavingsMsg'
Learn More

Diphenylmethane, 99%

Product Code. 10237110 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 10237110

Brand: Thermo Scientific Acros 117350010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 101-81-5 | C13H12 | 168.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-81-5
Molecular Formula C13H12
Molecular Weight (g/mol) 168.24
MDL Number MFCD00004781
InChI Key CZZYITDELCSZES-UHFFFAOYSA-N
Synonym diphenylmethane, ditan, ditane, benzene, 1,1'-methylenebis, diphenyl methane, benzene, benzyl, methane, diphenyl, 1,1'-dimethylenebis benzene, benzene, phenylmethyl, methylenedibenzene
PubChem CID 7580
ChEBI CHEBI:38884
IUPAC Name benzylbenzene
SMILES C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 24.5°C to 25.0°C
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 265.0°C
Flash Point 127°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2CH2
Refractive Index 1.5760 to 1.5780
Quantity 1 kg
Beilstein 05, II, 498
Merck Index 15, 3362
Specific Gravity 1
Solubility Information Solubility in water: practically insoluble. Other solubilities: insoluble in liquid ammonia, freely soluble in alcohol, ether, chloroform,, hexane and benzene
Formula Weight 168.24
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material Diphenylmethane, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.