missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylphosphine oxide, 97%, Thermo Scientific Chemicals
£42.40 - £130.00
Chemical Identifiers
CAS | 4559-70-0 |
---|---|
Molecular Formula | C12H11OP |
Molecular Weight (g/mol) | 202.19 |
MDL Number | MFCD00002079 |
InChI Key | ASUOLLHGALPRFK-UHFFFAOYSA-N |
Synonym | diphenylphosphine oxide, phosphine oxide, diphenyl, diphenyl phosphine oxide, hpoph2, asuollhgalprfk-uhfffaoysa-n, dppo, diphenylphosphane oxide, phenylphosphonoylbenzene, diphenylphosphino-1-one, pubchem23810 |
PubChem CID | 6327869 |
IUPAC Name | oxo(diphenyl)phosphanium |
SMILES | O=P(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10582091
|
Thermo Scientific Acros
310120010 |
1 g | Glass bottle |
£42.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10484575
|
Thermo Scientific Acros
310120050 |
5 g | Glass bottle |
£130.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4559-70-0 | |
202.19 | |
ASUOLLHGALPRFK-UHFFFAOYSA-N | |
6327869 | |
O=P(C1=CC=CC=C1)C1=CC=CC=C1 |
C12H11OP | |
MFCD00002079 | |
diphenylphosphine oxide, phosphine oxide, diphenyl, diphenyl phosphine oxide, hpoph2, asuollhgalprfk-uhfffaoysa-n, dppo, diphenylphosphane oxide, phenylphosphonoylbenzene, diphenylphosphino-1-one, pubchem23810 | |
oxo(diphenyl)phosphanium |
Specifications
4559-70-0 | |
Orange to Yellow | |
96% min. (GC) | |
(C6H5)2P(O)H | |
MFCD00002079 | |
ASUOLLHGALPRFK-UHFFFAOYSA-N | |
oxo(diphenyl)phosphanium | |
6327869 | |
97% | |
Diphenylphosphine oxide |
50.0°C to 57.0°C | |
Authentic | |
C12H11OP | |
1.6080 to 1.6100 | |
diphenylphosphine oxide, phosphine oxide, diphenyl, diphenyl phosphine oxide, hpoph2, asuollhgalprfk-uhfffaoysa-n, dppo, diphenylphosphane oxide, phenylphosphonoylbenzene, diphenylphosphino-1-one, pubchem23810 | |
O=P(C1=CC=CC=C1)C1=CC=CC=C1 | |
202.19 | |
202.2 | |
Crystals |
RUO – Research Use Only