Learn More
Diphenylphosphoryl azide, 98%, Thermo Scientific Chemicals
£50.95 - £682.40
Chemical Identifiers
CAS | 26386-88-9 |
---|---|
Molecular Formula | C12H10N3O3P |
Molecular Weight (g/mol) | 275.20 |
MDL Number | MFCD00001987 |
InChI Key | SORGEQQSQGNZFI-UHFFFAOYSA-N |
Synonym | diphenylphosphoryl azide, diphenyl azidophosphate, diphenylphosphonic azide, diphenyl phosphoryl azide, diphenyl phosphorazidate, phosphorazidic acid, diphenyl ester, azido phenoxy phosphoryl oxy benzene, dppa polymer-bound, diphenylphosphorazidate, unii-gxm91165av |
PubChem CID | 123414 |
IUPAC Name | [azido(phenoxy)phosphoryl]oxybenzene |
SMILES | [N-]=[N+]=NP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10686051
|
Thermo Scientific Acros
409040250 |
25 g | Glass bottle |
£50.95
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10178112
|
Thermo Scientific Acros
409041000 |
100 g | Glass bottle |
£146.73
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10649443
|
Thermo Scientific Acros
409045000 |
500 g | Glass bottle |
£682.40
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, For Azidation/Diazo transferIdentificadores de productos químicos
26386-88-9 | |
275.20 | |
SORGEQQSQGNZFI-UHFFFAOYSA-N | |
123414 | |
[N-]=[N+]=NP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1 |
C12H10N3O3P | |
MFCD00001987 | |
diphenylphosphoryl azide, diphenyl azidophosphate, diphenylphosphonic azide, diphenyl phosphoryl azide, diphenyl phosphorazidate, phosphorazidic acid, diphenyl ester, azido phenoxy phosphoryl oxy benzene, dppa polymer-bound, diphenylphosphorazidate, unii-gxm91165av | |
[azido(phenoxy)phosphoryl]oxybenzene |
Especificaciones
26386-88-9 | |
1.2730g/mL | |
112°C | |
97.5% min. (GC) | |
1.5500 to 1.5520 | |
04,210; 05,280; 07,138; 08,211; 10,173 | |
1.273 | |
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents | |
[N-]=[N+]=NP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1 | |
275.20 | |
275.19 | |
Liquid |
Yellow to Amber | |
157.0°C (0.2 mmHg) | |
Authentic | |
C12H10N3O3P | |
MFCD00001987 | |
15,3366 | |
diphenylphosphoryl azide, diphenyl azidophosphate, diphenylphosphonic azide, diphenyl phosphoryl azide, diphenyl phosphorazidate, phosphorazidic acid, diphenyl ester, azido phenoxy phosphoryl oxy benzene, dppa polymer-bound, diphenylphosphorazidate, unii-gxm91165av | |
SORGEQQSQGNZFI-UHFFFAOYSA-N | |
[azido(phenoxy)phosphoryl]oxybenzene | |
123414 | |
98% | |
Diphenylphosphoryl azide |
Seguridad y manipulación
GHS H Statement
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occur
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 247-644-
- Diphenylphosphoryl azide
Palabra de advertencia
- Danger
Clasificación
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Indicaciones de peligro
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
Consejos de prudencia
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only