Learn More
Diphenylsilanediol, 98%, Thermo Scientific Chemicals
£20.00 - £208.04
Chemical Identifiers
CAS | 947-42-2 |
---|---|
Molecular Formula | C12H12O2Si |
Molecular Weight (g/mol) | 216.311 |
MDL Number | MFCD00002101 |
InChI Key | OLLFKUHHDPMQFR-UHFFFAOYSA-N |
Synonym | diphenylsilanediol, silanediol, diphenyl, diphenyldihydroxysilane, difenyl-dihydroxysilan, silane, dihydroxydiphenyl, dihydroxy diphenyl silane, diphenylsilandiol, silanediol, 1,1-diphenyl, unii-36x37p8nbb, difenyl-dihydroxysilan czechShow More |
PubChem CID | 13693 |
IUPAC Name | dihydroxy(diphenyl)silane |
SMILES | C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(O)O |
Produktcode | Marke | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11494270
|
Thermo Scientific Alfa Aesar
A10492.14 |
25 g |
£20.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404280
|
Thermo Scientific Alfa Aesar
A10492.22 |
100 g |
£53.06
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414280
|
Thermo Scientific Alfa Aesar
A10492.36 |
500 g |
£208.04
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Diphenylsilanediol is used in Medicine field, electronics industry and polymer material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiphenylsilanediol is used in Medicine field, electronics industry and polymer material.
Solubility
Reacts with water.
Notes
Stable under normal temperatures and pressures. Incompatible with Oxidizing agents. Store at room temperature. Air & Light Sensitive.
Chemical Identifiers
947-42-2 | |
216.311 | |
OLLFKUHHDPMQFR-UHFFFAOYSA-N | |
13693 | |
C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(O)O |
C12H12O2Si | |
MFCD00002101 | |
diphenylsilanediol, silanediol, diphenyl, diphenyldihydroxysilane, difenyl-dihydroxysilan, silane, dihydroxydiphenyl, dihydroxy diphenyl silane, diphenylsilandiol, silanediol, 1,1-diphenyl, unii-36x37p8nbb, difenyl-dihydroxysilan czechShow More | |
dihydroxy(diphenyl)silane |
Specifications
∼150°C (decomposition) | |
C12H12O2Si | |
MFCD00002101 | |
2523445 | |
OLLFKUHHDPMQFR-UHFFFAOYSA-N | |
dihydroxy(diphenyl)silane | |
13693 | |
98% | |
Diphenylsilanediol |
947-42-2 | |
(C6H5Si(OH)2 | |
UN1325 | |
diphenylsilanediol, silanediol, diphenyl, diphenyldihydroxysilane, difenyl-dihydroxysilan, silane, dihydroxydiphenyl, dihydroxy diphenyl silane, diphenylsilandiol, silanediol, 1,1-diphenyl, unii-36x37p8nbb, difenyl-dihydroxysilan czech | |
C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(O)O | |
216.311 | |
216.31 | |
Odorless |
Safety and Handling
GHS H Statement
H228
Flammable solid.
GHS P Statement
P210
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
missing translation for 'rtecsNumber' : VV3640000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.