Learn More
Diphenyltin sulfide, 99%, Thermo Scientific™
£28.53 - £112.00
Chemical Identifiers
CAS | 20332-10-9 |
---|---|
Molecular Formula | C12H10SSn |
Molecular Weight (g/mol) | 304.982 |
MDL Number | MFCD00058856 |
InChI Key | CYXRBVIJBDYNQU-UHFFFAOYSA-N |
Synonym | diphenyltin sulfide, diphenylthioxostannane, diphenylstannanethione, diphenyl tin sulfide, diphenyl thioxo stannane, ph2sns, acmc-209ums, diphenyl sulfanylidene tin |
PubChem CID | 88492 |
IUPAC Name | diphenyl(sulfanylidene)tin |
SMILES | C1=CC=C(C=C1)[Sn](=S)C2=CC=CC=C2 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
20332-10-9 | |
304.982 | |
CYXRBVIJBDYNQU-UHFFFAOYSA-N | |
88492 | |
C1=CC=C(C=C1)[Sn](=S)C2=CC=CC=C2 |
C12H10SSn | |
MFCD00058856 | |
diphenyltin sulfide, diphenylthioxostannane, diphenylstannanethione, diphenyl tin sulfide, diphenyl thioxo stannane, ph2sns, acmc-209ums, diphenyl sulfanylidene tin | |
diphenyl(sulfanylidene)tin |
Specifications
20332-10-9 | |
MFCD00058856 | |
diphenyltin sulfide, diphenylthioxostannane, diphenylstannanethione, diphenyl tin sulfide, diphenyl thioxo stannane, ph2sns, acmc-209ums, diphenyl sulfanylidene tin | |
C1=CC=C(C=C1)[Sn](=S)C2=CC=CC=C2 | |
304.982 | |
304.97 | |
Powder |
C12H10SSn | |
UN3146 | |
CYXRBVIJBDYNQU-UHFFFAOYSA-N | |
diphenyl(sulfanylidene)tin | |
88492 | |
99% | |
Diphenyltin sulfide |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
missing translation for 'einecsNumber' : 243-739-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures