missing translation for 'onlineSavingsMsg'
Learn More

DIPPA hydrochloride, Tocris Bioscience™

Product Code. 30001435
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective and irreversible κ antagonist

An irreversible and selective antagonist at the κ receptor, with persistent effect in vivo.

Chemical Identifiers

CAS 155512-52-0
Molecular Formula C22H24Cl3N3OS
Molecular Weight (g/mol) 484.864
InChI Key BNWYENYHNOESCX-ZMBIFBSDSA-N
Synonym dippa hydrochloride, s-dippa hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-3-isothiocyanatophenyl-2-1-pyrrolidinyl ethyl acetamide hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-3-isothiocyanatophenyl-2-1-pyrrolidinyl ethyl acetamide hydrochloride
PubChem CID 45073425
IUPAC Name 2-(3,4-dichlorophenyl)-N-[(1S)-1-(3-isothiocyanatophenyl)-2-pyrrolidin-1-ylethyl]-N-methylacetamide;hydrochloride
SMILES CN(C(CN1CCCC1)C2=CC(=CC=C2)N=C=S)C(=O)CC3=CC(=C(C=C3)Cl)Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 484.87
Percent Purity >99%
Chemical Name or Material DIPPA hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'