missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipropylene glycol, 99%, mixture of isomers, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
3 kg
Packaging:
Glass Bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Note: The chemical structure above shows only one of the molecules included in this mixture.
Spécification
Spécification
Color | Orange-Red to Yellow |
Density | 1.0200g/mL |
Boiling Point | 229.0°C to 232.0°C |
Flash Point | 138°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Plastic drum |
Linear Formula | CH3CH(OH)CH2OCH2CH(OH)CH3 |
Refractive Index | 1.4400 to 1.4420 |
Quantity | 3 kg |
Afficher plus |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'