missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipropylene glycol, 99%, mixture of isomers, Thermo Scientific Chemicals
£28.40 - £60.20
Chemical Identifiers
CAS | 25265-71-8 |
---|---|
Molecular Formula | C6H14O3 |
Molecular Weight (g/mol) | 134.18 |
MDL Number | MFCD00051023 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10289550
|
Thermo Scientific Acros
409150010 |
1 kg | Glass Bottle |
£28.40
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10742212
|
Thermo Scientific Acros
409150030 |
3 kg | Plastic drum |
£60.20
3kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Note: The chemical structure above shows only one of the molecules included in this mixture.Chemical Identifiers
25265-71-8 | |
C6H14O3 | |
134.18 | |
MFCD00051023 |
Specifications
25265-71-8 | |
1.0200g/mL | |
138°C | |
98.5% min. (GC) | |
CH3CH(OH)CH2OCH2CH(OH)CH3 | |
MFCD00051023 | |
Solubility in water: miscible. Other solubilities: soluble in ethanol | |
134.18 | |
Liquid |
Orange-Red to Yellow | |
229.0°C to 232.0°C | |
Authentic | |
C6H14O3 | |
1.4400 to 1.4420 | |
1.02 | |
134.18 | |
99% | |
Dipropylene glycol, 99% |
Safety and Handling
missing translation for 'einecsNumber' : 246-770-3
missing translation for 'rtecsNumber' : UB8765000
missing translation for 'tsca' : TSCA
RUO – Research Use Only