missing translation for 'onlineSavingsMsg'
Learn More

Dipropylene glycol, mixture of isomers, 99%, Thermo Scientific Chemicals

Product Code. 11492988
Click to view available options
Quantity:
500 g
2500 g

Dipropylene glycol, mixture of isomers is used as a solvent, plasticizer, polymerization initiator and monomer. It acts as an additive used in perfumes, skin and hair care products. It is an active ingredient in commercial fog fluid, which is used as an entertainment in fog machines. Further, it is used as an intermediate in polyurethane polyols, alkyd resins and non-reinforced polyesters. In addition to this, it is used in dyes, paints, hydraulic brake fluids and cutting oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dipropylene glycol, mixture of isomers is used as a solvent, plasticizer, polymerization initiator and monomer. It acts as an additive used in perfumes, skin and hair care products. It is an active ingredient in commercial fog fluid, which is used as an entertainment in fog machines. Further, it is used as an intermediate in polyurethane polyols, alkyd resins and non-reinforced polyesters. In addition to this, it is used in dyes, paints, hydraulic brake fluids and cutting oils.

Solubility
Miscible with cold water, hot water, methanol, diethyl ether and ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25265-71-8
Molecular Weight (g/mol) 134.18
InChI Key AZUXKVXMJOIAOF-UHFFFAOYNA-N
PubChem CID 8087
IUPAC Name 1-(2-hydroxypropoxy)propan-2-ol
SMILES CC(O)COCC(C)O

Specifications

Melting Point °C to 32°C
Density 1.023
Boiling Point 231°C to 235°C
Assay Percent Range 0.99
Molecular Formula C6H14O3
Refractive Index 1.441
MDL Number MFCD00004538
Quantity 500 g
Synonym 1,1'-oxydi-2-propanol, 2-propanol, 1,1'-oxybis, 1,1'-oxydipropan-2-ol, bis 2-hydroxypropyl ether, 4-oxaheptane-2,6-diol, 1,1'-oxybis-2-propanol, 2,2'-dihydroxydipropyl ether, 2-propanol, 1,1'-oxydi, niax catalyst d-19, 1,1'-dimethyldiethylene glycol
Solubility Information Miscible with cold water,hot water,methanol,diethyl ether and ethanol.
Formula Weight 134.18
Chemical Name or Material Dipropylene glycol, mixture of isomers
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'