missing translation for 'onlineSavingsMsg'
Learn More

Microspec™ DipSlides

For semi-quantitative microbiological testing.

£26.90 - £28.50

Specifications

ATCC® Yes
Cap Color White
Cap Type Plug
Color Clear
Content And Storage 2°C to 8°C
View More Specs

Products 2
Product Code Brand Additive Price Quantity & Availability  
Product Code Brand Additive Price Quantity & Availability  
15712145
View Documents
Microspec™
O1
TTC/MEA
£26.90
Pack of 10
In Stock
to see stock.
Edge
Add to basket
 
15722145
View Documents
Microspec™
O2
TTC/TTC
£28.50
Pack of 10
Estimated Shipment: 23-07-2025
to see stock.
Add to basket
 
Description

Description

  • Used by non-skilled field personnel
  • Comes with easy to interpret results chart
  • Hinged handle design
  • Ideal for water testing such as cooling tower monitoring
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 6 Months

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Microspec™ DipSlides

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.