missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dirhenium decacarbonyl, sublimed, Thermo Scientific Chemicals
Click to view available options
5g; Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 14285-68-8 |
Melting Point | 170.0°C |
Color | Green-Yellow |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Molecular Formula | C10O10Re2 |
Linear Formula | Re2(CO)10 |
MDL Number | MFCD00011198 |
Quantity | 5 g |
Synonym | carbon monoxide; rhenium 2+, decacarbonyldirhenium 250mg |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Dirhenium decacarbonyl, sublimed
- Warning
- Acute toxicity Category 4
- H332-Harmful if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'