missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Comparator Disc
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
Brand: Lovibond™ 221130
This item is not returnable.
View return policy
Description
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
- Easy to use, visual assessment of the sample; simply rotate disc to match color
- Glass, fade-free standards for longevity of use
- Shock resistant, scratch resistant
- Optional lighting units for guaranteed lighting conditions 24 hours/day
- Optional Nessleriser systems for longer path lengths
- More than 400 color discs are available for the Comparator 2000+
- Lovibond™ color standards consist exclusively of through-and-through colored and flat-ground glasses
Specifications
ph Value | |
Comparator 2000+ | |
Color Comparator Disc |
Disc 2/1J | |
6.8, 7, 7.2, 7.4, 7.6, 7.8, 8, 8.2, 8.4 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lovibond™ Comparator Disc > Analyte: ph Value
Spot an opportunity for improvement?Share a Content Correction