missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Comparator Disc
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
Brand: Lovibond™ 221030
This item is not returnable.
View return policy
Beschreibung
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
- Easy to use, visual assessment of the sample; simply rotate disc to match color
- Glass, fade-free standards for longevity of use
- Shock resistant, scratch resistant
- Optional lighting units for guaranteed lighting conditions 24 hours/day
- Optional Nessleriser systems for longer path lengths
- More than 400 color discs are available for the Comparator 2000+
- Lovibond™ color standards consist exclusively of through-and-through colored and flat-ground glasses
Spezifikation
ph Value | |
Comparator 2000+ | |
Color Comparator Disc |
Disc 2/1B | |
2.8, 3, 3.2, 3.4, 3.6, 3.8, 4, 4.2, 4.4 |