Learn More
Lovibond™ Comparator Disc
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
Brand: Lovibond™ 221110
Code : 03
Additional Details : Weight : 1.00000kg
Description
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
- Easy to use, visual assessment of the sample; simply rotate disc to match color
- Glass, fade-free standards for longevity of use
- Shock resistant, scratch resistant
- Optional lighting units for guaranteed lighting conditions 24 hours/day
- Optional Nessleriser systems for longer path lengths
- More than 400 color discs are available for the Comparator 2000+
- Lovibond™ color standards consist exclusively of through-and-through colored and flat-ground glasses
Specifications
ph Value | |
Comparator 2000+ | |
Color Comparator Disc |
Disc 2/1H | |
6, 6.2, 6.4, 6.6, 6.8, 7, 7.2, 7.4, 7.6 |