Learn More
Cole-Parmer™ Stuart™ Discharge Tube for Pressmatic Bottle Top Dispenser
Choose a discharge tube for Model PD10P or PD60P Pressmatic Bottle Top Dispenser
Brand:  Cole-Parmer™ 99955-60
 View more versions of this product
Product Code. 12437772
  • £41.95 / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with a Stuart™ Model PD10P or PD60P Pressmatic Bottle Top Dispensers
  • PD10P works with smaller volumes from 2 to 10mL
  • PD60P works with volumes between 10 and 60mL
  • Compatible with dilute acids, buffers, polar solvents, biological solutions
Specifications
Specifications
Discharge Tube
Stuart PD10P Pressmatic