Learn More
BRAND™ Discharge Valves for Dispensette™ S Bottletop Dispensers
Valves for Dispensette™ S / S Organic / S Trace Analysis
Brand:  BRAND™ 6729
27.50 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15653977
  • £38.85 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Discharge valves are manufactured of PFA/Boro 3.3/ceramic components. The Dispensette™ S discharge valve has a platinum-iridium valve spring; the Dispensette™ S Organic discharge valve has a tantalum valve spring.
  • Discharge valves for the Dispensette™ S Trace Analysis are available with either a platinum-iridium or tantalum valve spring for specific applications (e.g., Pt-Ir valve for dispensing HF). Available in two or three sizes: one for 1mL and 2mL models; one for 5mL and 10mL models; one for 25mL, 50mL and 100mL models
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Discharge Valve
Translucent
Dispensing liquid
Discharge Valve Only
Autoclavable
ETFE, Borosilicate Glass, Ceramic
Dispensette™ S Organic, 5 and 10 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BRAND™ Discharge Valves for Dispensette™ S Bottletop Dispensers > For Dispensette S Organic 5&10mL nominal capacity