missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Day Impex™ Virkon™ Disinfectant Virucidal Tablets
Clean and disinfect work surfaces and equipment in a single, one-step operation. Day Impex™ Virkon™ Disinfectant Virucidal Tablets provide the widest proven spectrum of any disinfectant.
£282.00 - £724.00
Specifications
Product Type | Virucidal Disinfectant |
---|---|
Form | Tablet |
For Use With (Application) | For Cleaning and Disinfecting Work Surfaces and Equipment |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12318667
|
Day Impex™
330013 |
50 Tablets/Tub, 12 Tubs/Cs. |
£724.00
600 tablets |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12328667
|
Day Impex™
330012 |
10 Tablets/Tub, 20 Tubs/Cs. |
£282.00
200 tablets |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Use biocides safely. Always read the label and product information before use.
- Widest proven spectrum of any disinfectant - proven effective in independent tests against all 18 virus families affecting man and animals, including AIDS (HIV) and Hepatitis B
- Excellent bactericide and fungicide - cleans and disinfects work surfaces and equipment in a single, one-step operation
- Can be applied in powder form direct to spillages of blood and other body fluids
- Easily dissolves to make a liquid disinfectant
- Odourless, non-irritant to skin and eyes, non-toxic vapour phase, exceptionally low oral toxicity
- Under COSHH requirements, no maximum exposure limit is required
- Non-corrosive, non-bleaching and biodegradable
- Any viral disease transmitted via surfaces or instruments contaminated with infected blood, body fluids, faeces or saliva can be controlled by correct application For cleaning and disinfecting work surfaces and equipment.
Specifications
Virucidal Disinfectant | |
Tablet | |
For Cleaning and Disinfecting Work Surfaces and Equipment |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- relyon virkon tablets
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P264-Wash skin thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : pentapotassium bis(peroxymonosulphate) bis(sulphate), sulphamidic acid, benzenesulphonic acid, potas
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Day Impex™ Virkon™ Disinfectant Virucidal Tablets