missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Disitertide
TGF-β1 inhibitor
Brand: Tocris Bioscience™ 7717/1
Additional Details : Weight : 0.00970kg
Description
Disitertide is a transforming growth factor-beta 1 (TGF-β1) inhibitor. In vitro, it inactivates TGF-β1/Smads signaling which reduces cell migration and invasion without affecting cell proliferation in colorectal carcinoma cell lines (Lovo and SW480 cells).Specifications
7040 | |
272105-42-7 | |
CEDLAP, DPD1, latency-associated peptide, TGF beta, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 | |
1 mg |
0.95 | |
Store at -20°C | |
M.W.-observed: 1580.83 |