missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disodium terephthalate, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Description
Disodium terephthalate and its various derivatives are synthesized via simple acid-base chemistry for anode materials in Na ion batteries. They show excellent electrochemical performance, including little capacity fading over 90 cycles, ideal redox potential, and excellent rate performance, making them promising candidates for Na ion batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Quantity | 500 g |
Solubility Information | Soluble in water. |
Formula Weight | 210.1 |
Percent Purity | ≥99% |
Physical Form | Powder |
Chemical Name or Material | Disodium terephthalate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'