Learn More
Fisherbrand™ Dispenser tips
Robust, stable, and durable dispenser tips for several brands of pipettors. For dispensing of fluids with high viscosity and high vapor pressure.
Brand:  Fisherbrand™ 702628
149.18 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
 View more versions of this product
Product Code. 15133656
  • £262.00 / Pack of 100
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • These disposable tips are suitable for the following multiple dispensers:
  • BRAND Handystep™ S, HandyStep™ electronic, HandyStep™ touch/ S
  • Eppendorf Multipette™ 4780
  • Gilson Distriman™ and Gilson Repetman™
  • Ritter Ripette™ pro
  • EDOS™ 5221
  • Rainin AutoRep™ E, Rainin AutoRep™ S, Rainin AutoRep™ M
  • STARLAB RepeatOne™
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2.5 mL
Polypropylene
For dispensing of fluids with high viscosity, high vapor pressure
Single-use
Sterile
ISO 8655
Dispenser Tip
Polypropylene