Learn More
BRAND™ Dispensette™ S Analog-adjustable Bottletop Dispensers
GREENER_CHOICE
The Dispensette™ S Analog Bottletop Dispensers feature a floating piston design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes. Designed for general-purpose dispensing applications.
£388.00 - £937.00
Specifications
Product Type Bottletop Dispenser
Autoclavable Autoclavable (121°C)
Certifications/Compliance ISO 9001
Color Red
Display Analog
View More Specs
Includes: Discharge tube, valve mounting/calibration tool, polypropylene adapters, telescoping filling tube, recirculation tube (optional), performance certificate, operating manual
Products 14
Product Code Brand Volume (Metric) Accuracy Description Increments Price Quantity & Availability  
Product Code Brand Volume (Metric) Accuracy Description Increments Price Quantity & Availability  
15478043
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4600101
0.1 to 1 mL 0.6% (6μL) With recirculation valve 0.02 mL
£446.00
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
15498043
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4600121
0.2 to 2 mL 0.5% (10μL) With recirculation valve 0.05 mL
£496.00
Each
Estimated Shipment: 01-05-2025
to see stock.
Add to basket
15429032
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4600131
0.5 to 5 mL 0.5% (25μL) With recirculation valve 0.1 mL
£496.00
Each
In Stock
to see stock.
Add to basket
15444952
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4600141
1 to 10 mL 0.5% (50μL) With recirculation valve 0.2 mL
£512.00
Each
In Stock
to see stock.
Edge
Add to basket
15454942
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4600151
2.5 to 25 mL 0.5% (125μL) With recirculation valve 0.5 mL
£621.00
Each
In Stock
to see stock.
Add to basket
15406932
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4600161
5 to 50 mL 0.5% (250μL) With recirculation valve 1.0 mL
£715.00
Each
In Stock
to see stock.
Edge
Add to basket
15417361
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4600171
10 to 100 mL 0.5% (500μL) With recirculation valve 1.0 mL
£937.00
Each
In Stock
to see stock.
Add to basket
15488043
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4600120
0.2 to 2 mL 0.5% (10μL) Without recirculation valve 0.05 mL
£434.00
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
15468043
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4600100
0.1 to 1 mL 0.6% (6μL) Without recirculation valve 0.02 mL
£388.00
Each
In Stock
to see stock.
Add to basket
15408053
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4600130
0.5 to 5 mL 0.5% (25μL) Without recirculation valve 0.1 mL
£400.00
Each
In Stock
to see stock.
Add to basket
15418053
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4600140
1 to 10 mL 0.5% (50μL) Without recirculation valve 0.2 mL
£445.00
Each
In Stock
to see stock.
Add to basket
15428053
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4600150
2.5 to 25 mL 0.5% (125μL) Without recirculation valve 0.5 mL
£506.00
Each
In Stock
to see stock.
Add to basket
15438053
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4600160
5 to 50 mL 0.5% (250μL) Without recirculation valve 1.0 mL
£638.00
Each
In Stock
to see stock.
Add to basket
15492823
GREENER_CHOICE
View Documents Promotion Details
BRAND™
4600170
10 to 100 mL 0.5% (500μL) Without recirculation valve 1.0 mL
£823.00
Each
In Stock
to see stock.
Add to basket
Description
Description
  • New discharge tube, with or without recirculation valve
  • New valve system - no additional sealing rings required, facilitates cleaning and preparations for autoclaving
  • Simple priming by optimized flow channels
  • Less dispensing force - especially with large volume instruments
  • Safe volume setting with internal scalloped track (analog-adjustable instruments)
  • Pivoted screw cap - does not disturb dispensing
  • Easy to calibrate and adjust in order to comply with GLP guidelines
  • Easy to dismantle for cleaning
  • Replacable filling and discharge valve with safety bulb (locks when discharge tube is not assembled)
  • 360° rotatable valve block allows bottle label to face user
  • Telescoping filling tube adjusts easily to different size bottles
  • DE-M marking
Warranty
One year
Specifications
Specifications
Bottletop Dispenser
ISO 9001
Analog
Autoclavable (121°C)
Red
Dispensing aggressive reagents, including concentrated bases and acids such as H3PO4, H2SO4 (except e.g., HCl, HNO3 und HF), saline solutions, as well as many organic solvents
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
BRAND™ Dispensette™ S Analog-adjustable Bottletop Dispensers