Learn More
Kinematica Dispersing Vessel
Cloverleaf internal shape effectively interrupts the flow in the radial direction and generates additional counteracting force.
Brand:  Kinematica 11051041
Product Code. 15883744
  • £160.00 / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cloverleaf internal shape
  • Interrupts flow in radial direction to generate additional counteracting forces
  • High turbulences maximize mixing and dispersion efficiency
  • Minimizes processing time and energy output
Specifications
Specifications
150 mL
20 mm
150 mm
Thread: GL45
54 mm
GS 25KL2