Learn More
CellPath DispoCut™ Board
Disposable cutting board designed to increase safety and hygiene in high-risk areas as well as routine cutting areas.
Brand:  CellPath CGB-0502-53A
Product Code. 12826653
  • £208.00 / Pack of 48
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from 3mm, three ply foam filled board for strength and rigidity
  • Offers high degree of moisture resistance and helps protect work surfaces while ensuring accurate dissection
  • Features metric and imperial scale to assist in macroscopic and photographic recording of specimens
  • Easily disposed of by incineration
Specifications
Specifications
White
148 x 210 mm