Learn More
Epredia™ Disposable Base Molds
Product Code. p-4524715 Shop All Epredia Products
Click to view available options
Dimensions (L x W):
15 x 15 mm
24 x 24 mm
30 x 24 mm
37 x 24 mm
Unit Size:
Pack of 500
Product Code. 12590107
Brand: Epredia™ 58950
Use Epredia™ Disposable Base Molds with all standard-size cassettes.
  • Cleaning associated with metal molds is not required
  • Transparency facilitates tissue orientation
Ordering Information:
  • Sold in cases of four boxes.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Base Mold
Material Plastic
Dimensions (L x W) 15 x 15 mm
Disposable Single-use