Learn More
Product Code | Brand | Product Type | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | For Use With (Equipment) | Price | Quantity & Availability | |||||
12647876
|
Epredia™
705050 |
Disposable Blade Carrier E | Dovetail Style Unit | N/A | ||||||
12033119
|
Epredia™
705800 |
Blade Carrier E | HM 310, HM 315 and HM 325 |
£1390.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Disposable Blade Carriers- Blade angle and blade adjustment levers are removable to help protect user settings (Version 2 and 3 units only)
- Concentric lever locks onto the dovetail to help reduce accidental movement of the holder (Version 2 and 3 units only)
- Quick clamp system for high- and low-profile blades
- Adjustable clearance angle
- Moveable knife guard covers the blade entirely in any position
- Knife carrier base included
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Epredia™ Disposable Blade Carriers