missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Disposable Healthcare Tunic and Pants Set, 48 g/m2

Product Code. 17750169
missing translation for 'orderingAttributeHoverText'
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

These sets of disposable tunics and pants provide extra protection for healthcare workers.

  • Light and breathable polypropylene material
  • Loose shape allows user to maintain good movement
  • Collarless shirt
  • Pants with elasticated waist
  • Individual packaging for hygiene and controlled distribution
  • 48 g/m2 fabric weight

Specifications

Size Small
Material Polypropylene
Color Navy
Sleeve Style Short Sleeve
Disposable Single-use
For Use With (Application) Dental, Medical
No. per Pack 60 Pk.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'