missing translation for 'onlineSavingsMsg'
Learn More
Kolmi Hopen™ Disposable Healthcare Tunic and Pants Set, 48 g/m2 Product Code.: 17770169
Estimated Shipment: 25-04-2025
to see stock.
Add to basket

Kolmi Hopen™ Disposable Healthcare Tunic and Pants Set, 48 g/m2

Product Code. 17770169
2X-Large
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy

Product Code. 17770169

Brand: Kolmi Hopen™ J40480/XXL

Estimated Shipment: 25-04-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

These sets of disposable tunics and pants provide extra protection for healthcare workers.

  • Light and breathable polypropylene material
  • Loose shape allows user to maintain good movement
  • Collarless shirt
  • Pants with elasticated waist
  • Individual packaging for hygiene and controlled distribution
  • 48 g/m2 fabric weight

Specifications

Size 2X-Large
Material Polypropylene
Color Navy
Sleeve Style Short Sleeve
Disposable Single-use
For Use With (Application) Dental, Medical
No. per Pack 60 Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kolmi Hopen™ Disposable Healthcare Tunic and Pants Set, 48 g/m2 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.