missing translation for 'onlineSavingsMsg'
Learn More

Disposable Inoculation Loop, 10μL

Manufactured from polystyrene for strength and flexibility. Microspec™ Disposable Inoculation Loops are ideal for the cultivation of microbes on plates by transferring inoculum for streaking and to transfer microscopic organisms.

Brand:  Microspec A3

Product Code. 12870155

  • £148.00 / Pack of 1000
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • High impact, polystyrene, disposable inoculating loops
  • 200mm long to reduce the risk of contamination
  • Aseptic Manufacture
Specifications

Specifications

Single-use
200 mm
Aseptically Manufactured
Laboratory and Research use
Laboratory and Research use
10 μL
Plastic
Inoculating Loop
1000 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Disposable Inoculation Loop, 10μL > Volume: 10uL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.