missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Disposable IR Cards, 5x10cm, NaCl window, 19mm aperture

Product Code. p-7070057
Click to view available options
Quantity:
25 Pk.
5 Pk.
Unit Size:
Pack of 25
Pack of 5
This item is not returnable. View return policy

Product Code. 15416565

Brand: Thermo Scientific Chemicals 043929.KM

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Size: 5 x 10cm
  • NaCl window
  • 19mm aperture
TRUSTED_SUSTAINABILITY

Specifications

Description Disposable IR Cards
Quantity 25 Pk.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.