Learn More
Disposable Luer-Slip Two-Part Tuberculin Syringe
Safe application of medication is ensured by the high quality of these needles. Henke Sass Wolf Hsw™ Disposable Luer-Slip Two-Part Tuberculin Syringes are designed for optimized user handling to ensure patient safety.
Brand:  Normject 4010.200V0
Product Code. 10795874
  • £11.10 / Pack of 100
Edge
Explore more special offers
Includes: Tuberculin Syringes only. (Needles not included; sold separately.)
Description
Description
  • Fully visible glass barrel for visual verification
  • Ergonomic light weight metal, plastic design
  • 2mlL barrel with 0.1mL increment trigger
  • Safe and easy needle change out via luer lock connection
  • Limited spare parts for easy service
  • Boilable
Note:  Needles are sold separately.
Specifications
Specifications
Autoclavable
Polypropylene
Smudge-proof scale
Yes
Single-use
100/Pk.
Leur Slip
Sterile
DIN, EN, ISO and FDA standards, Quality management system certified according to ISO 13485:2003 / AC 2007, CE-marked
Transparent Barrel
Syringes only. (Needles are sold separately.)
Luer-slip 2-part Tuberculin Syringe
0.01 mL
1 mL
Yes