missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Disposable Magnetic Stirring Bars
Stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
£180.00 - £233.47
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | ISO 9001:2008 |
Diameter (Metric) | 8 mm |
Shape | Polygon |
For Use With (Application) | Mixing radioactive reagents when cross contamination might be a concern |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
11764146
|
Bel-Art™ SP Scienceware™
F37112-0002 |
50.8 mm |
£233.47
Pack of 100 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
11754146
|
Bel-Art™ SP Scienceware™
F37112-0016 |
41.3 mm |
£180.00
Pack of 100 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
Description
- Useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
- Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
- Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
- Autoclave safe
Specifications
Autoclavable | |
8 mm | |
Mixing radioactive reagents when cross contamination might be a concern |
ISO 9001:2008 | |
Polygon |